siRNA Details
Gene Symbol: VPS39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301138.1 | NP_001288067.1 | 14 | 1454 | ||
NM_015289.3 | NP_056104.2 | 13 | 1421 | |||
XM_011521403.2 | XP_011519705.1 | 14 | 1423 | |||
XM_011521404.2 | XP_011519706.1 | 13 | 1426 | |||
XM_017022036.1 | XP_016877525.1 | 14 | 1422 | |||
XM_017022037.1 | XP_016877526.1 | 13 | 1426 | |||
XM_017022038.1 | XP_016877527.1 | 13 | 1609 | |||
XM_017022039.1 | XP_016877528.1 | 13 | 1420 | |||
GenBank mRNA | AB018313.2 | - | 13 | 1372 | ||
AF280814.1 | - | 14 | 1281 | |||
AF281052.1 | - | 13 | 1391 | |||
BC015817.2 | - | 9 | 981 | |||
BC068559.1 | - | 13 | 1390 | 5295166 | ||
BX537404.1 | - | 13 | 1400 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
