siRNA Details
Gene Symbol: GTF3C5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122823.1 | NP_001116295.1 | 6 | 1312 | ||
NM_001286709.1 | NP_001273638.1 | 4 | 893 | |||
NM_012087.3 | NP_036219.2 | 6 | 1312 | |||
XM_005272234.3 | XP_005272291.1 | 6 | 1307 | |||
XM_011519201.1 | XP_011517503.1 | 6 | 1040 | |||
XM_017015315.1 | XP_016870804.1 | 6 | 1040 | |||
XM_017015316.1 | XP_016870805.1 | 6 | 1040 | |||
GenBank mRNA | AF133124.1 | - | 6 | 1267 | ||
AJ420443.1 | - | 6 | 980 | |||
AK000194.1 | - | 5 | 781 | |||
AK024510.1 | - | 4 | 575 | |||
AK055092.1 | - | 6 | 1274 | |||
AK097295.1 | - | 6 | 957 | |||
AK225761.1 | - | 6 | 1279 | |||
AK293980.1 | - | 5 | 767 | |||
AY007123.1 | - | 6 | 976 | |||
BC009741.2 | - | 5 | 766 | |||
BC011355.1 | - | 6 | 1256 | 4157684 | ||
BC017337.2 | - | 6 | 1235 | 4872395 | ||
BC030157.2 | - | 6 | 970 | |||
BQ673969.1 | - | 4 | 547 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
