siRNA Details
Gene Symbol: GTF3C5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122823.1 | NP_001116295.1 | 10 | 1570 | ||
NM_001286709.1 | NP_001273638.1 | 7 | 1130 | |||
NM_012087.3 | NP_036219.2 | 9 | 1549 | |||
XM_017015315.1 | XP_016870804.1 | 10 | 1298 | |||
XM_017015316.1 | XP_016870805.1 | 9 | 1277 | |||
GenBank mRNA | AF133124.1 | - | 9 | 1504 | ||
AJ420443.1 | - | 10 | 1238 | |||
AK000194.1 | - | 9 | 1039 | |||
AK024510.1 | - | 7 | 812 | |||
AK055092.1 | - | 9 | 1511 | |||
AK097295.1 | - | 9 | 1194 | |||
AK225761.1 | - | 9 | 1516 | |||
AY007123.1 | - | 9 | 1213 | |||
BC009741.2 | - | 8 | 1003 | |||
BC011355.1 | - | 9 | 1493 | 4157684 | ||
BC017337.2 | - | 9 | 1472 | 4872395 | ||
BC030157.2 | - | 9 | 1207 | |||
BQ673969.1 | - | 7 | 786 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
