siRNA Details
Gene Symbol: NXPH4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007224.3 | NP_009155.1 | 2 | 259 | ||
XM_017018747.1 | XP_016874236.1 | 2 | 197 | |||
GenBank mRNA | AK094231.1 | - | 3 | 464 | ||
AK290949.1 | - | 2 | 197 | |||
AY358614.1 | - | 2 | 250 | |||
BC036679.1 | - | 2 | 253 | 5264190 | ||
BC053581.1 | - | 2 | 183 | 5174266 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
