siRNA Details
Gene Symbol: NXPH4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007224.3 | NP_009155.1 | 2 | 635 | ||
GenBank mRNA | AK094231.1 | - | 3 | 840 | ||
AK290949.1 | - | 2 | 573 | |||
AY358614.1 | - | 2 | 626 | |||
BC036679.1 | - | 2 | 629 | 5264190 | ||
BC053581.1 | - | 2 | 559 | 5174266 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
