siRNA Details
Gene Symbol: OR9A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001658.1 | NP_001001658.1 | 1 | 38 | ||
GenBank mRNA | BC137048.1 | - | 1 | 101 | 9021045 | |
BC137049.1 | - | 1 | 101 | 9021046 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
