siRNA Details
Gene Symbol: DNAH12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291661.1 | NP_001278590.1 | 64 | 10369 | ||
XM_011533466.1 | XP_011531768.1 | 64 | 10354 | |||
XM_011533467.1 | XP_011531769.1 | 64 | 10321 | |||
XM_011533468.1 | XP_011531770.1 | 64 | 10291 | |||
XM_011533469.1 | XP_011531771.1 | 63 | 10213 | |||
XM_017005860.1 | XP_016861349.1 | 64 | 10354 | |||
XM_017005861.1 | XP_016861350.1 | 63 | 11037 | |||
XM_017005862.1 | XP_016861351.1 | 43 | 6929 | |||
GenBank mRNA | AK097746.1 | - | 5 | 655 | ||
AK128592.1 | - | 12 | 1795 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
