siRNA Details
Gene Symbol: DNAH12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291661.1 | NP_001278590.1 | 72, 73 | 11817 | ||
XM_011533466.1 | XP_011531768.1 | 72, 73 | 11802 | |||
XM_011533467.1 | XP_011531769.1 | 72, 73 | 11769 | |||
XM_011533468.1 | XP_011531770.1 | 72, 73 | 11739 | |||
XM_011533469.1 | XP_011531771.1 | 71, 72 | 11661 | |||
XM_017005861.1 | XP_016861350.1 | 71, 72 | 12485 | |||
XM_017005862.1 | XP_016861351.1 | 51, 52 | 8377 | |||
GenBank mRNA | AK097746.1 | - | 12, 13 | 1965 | ||
AK128592.1 | - | 20, 21 | 3243 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
