siRNA Details
Gene Symbol: TOR3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022371.3 | NP_071766.2 | 3 | 780 | ||
GenBank mRNA | AJ299403.1 | - | 3 | 669 | ||
AJ299441.1 | - | 3 | 669 | |||
AK025998.1 | - | 3 | 659 | |||
AK130132.1 | - | 1 | 99 | |||
BC001085.1 | - | 3 | 678 | 2959819 | ||
BC007571.1 | - | 3 | 669 | 3029304 | ||
BC011746.1 | - | 3 | 665 | 3343691 | ||
BC018292.1 | - | 3 | 670 | 3457259 | ||
CR457354.1 | - | 3 | 628 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
