siRNA Details
Gene Symbol: TOR3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022371.3 | NP_071766.2 | 1 | 253 | ||
GenBank mRNA | AJ299403.1 | - | 1 | 142 | ||
AJ299441.1 | - | 1 | 142 | |||
AK025998.1 | - | 1 | 132 | |||
AK299966.1 | - | 1 | 116 | |||
AK316530.1 | - | 1 | 128 | |||
BC001085.1 | - | 1 | 151 | 2959819 | ||
BC007571.1 | - | 1 | 142 | 3029304 | ||
BC011746.1 | - | 1 | 138 | 3343691 | ||
BC018292.1 | - | 1 | 143 | 3457259 | ||
BC088368.1 | - | 1 | 146 | |||
CR457354.1 | - | 1 | 101 | |||
DA942161.1 | - | 1 | 253 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
