siRNA Details
Gene Symbol: PID1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100818.1 | NP_001094288.1 | 3 | 591 | ||
NM_017933.4 | NP_060403.3 | 4 | 802 | |||
XM_011511384.2 | XP_011509686.1 | 3 | 474 | |||
XM_017004404.1 | XP_016859893.1 | 4 | 5733 | |||
XM_017004405.1 | XP_016859894.1 | 5 | 672 | |||
XM_017004406.1 | XP_016859895.1 | 3 | 330 | |||
XM_017004407.1 | XP_016859896.1 | 2 | 387 | |||
GenBank mRNA | AB075874.1 | - | 4 | 457 | ||
AK000708.1 | - | 3 | 577 | |||
AK096636.1 | - | 3 | 533 | |||
AK125359.1 | - | 4 | 726 | |||
AY317148.1 | - | 3 | 423 | |||
BC040164.1 | - | 2 | 378 | 5589172 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
