siRNA Details
Gene Symbol: SOS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB209277.1 | - | 5, 6 | 815 | ||
BC117261.1 | - | 5, 6 | 751 | 40125812 | ||
BC143366.1 | - | 5, 6 | 751 | 9051871 | ||
BC143367.1 | - | 5, 6 | 751 | 9051872 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
