siRNA Details
Gene Symbol: ARID1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017519.2 | NP_059989.2 | 7 | 2468 | ||
NM_020732.3 | NP_065783.3 | 8 | 2507 | |||
XM_005267069.4 | XP_005267126.2 | 7 | 2543 | |||
XM_011535984.2 | XP_011534286.2 | 9 | 2625 | |||
XM_011535988.2 | XP_011534290.1 | 1 | 25 | |||
XM_017011103.1 | XP_016866592.1 | 9 | 2625 | |||
XM_017011104.1 | XP_016866593.1 | 9 | 2624 | |||
XM_017011105.1 | XP_016866594.1 | 9 | 2625 | |||
XM_017011106.1 | XP_016866595.1 | 9 | 2624 | |||
XM_017011108.1 | XP_016866597.1 | 9 | 2632 | |||
XM_017011109.1 | XP_016866598.1 | 5 | 4981 | |||
GenBank mRNA | AB033061.1 | - | 1 | 56 | ||
AF253515.1 | - | 7 | 1662 | |||
AF259792.1 | - | 3 | 410 | |||
AF468300.1 | - | 8 | 1037 | |||
AF521671.1 | - | 7 | 2255 | |||
AK025945.1 | - | 5 | 734 | |||
AK074180.1 | - | 5 | 618 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
