siRNA Details
Gene Symbol: RBMS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002897.4 | NP_002888.1 | 14 | 1741 | ||
NM_016836.3 | NP_058520.1 | 14 | 1750 | |||
XM_005246737.1 | XP_005246794.1 | 15 | 1509 | |||
XM_005246738.2 | XP_005246795.1 | 15 | 1844 | |||
XM_005246739.2 | XP_005246796.1 | 15 | 1714 | |||
XM_005246740.1 | XP_005246797.1 | 15 | 1500 | |||
XM_005246741.3 | XP_005246798.1 | 15 | 1711 | |||
XM_017004626.1 | XP_016860115.1 | 14 | 1787 | |||
GenBank mRNA | AK127157.1 | - | 5 | 2142 | ||
AK225887.1 | - | 15 | 1409 | |||
AK297812.1 | - | 15 | 1500 | |||
BC012992.2 | - | 14 | 1436 | 3530897 | ||
BC012993.2 | - | 14 | 1436 | 3530898 | ||
BC018951.2 | - | 14 | 1737 | 4137722 | ||
BC051889.1 | - | 14 | 1323 | |||
BX648246.1 | - | 7 | 8907 | |||
L11289.1 | - | 14 | 1373 | |||
X64652.1 | - | 13 | 1256 | |||
X77494.1 | - | 16 | 1486 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
