siRNA Details
Gene Symbol: RBMS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002897.4 | NP_002888.1 | 14 | 1792 | ||
NM_016836.3 | NP_058520.1 | 14 | 1801 | |||
XM_005246737.1 | XP_005246794.1 | 15 | 1560 | |||
XM_005246738.2 | XP_005246795.1 | 15 | 1895 | |||
XM_005246739.2 | XP_005246796.1 | 15 | 1765 | |||
XM_005246740.1 | XP_005246797.1 | 15 | 1551 | |||
XM_005246741.3 | XP_005246798.1 | 15 | 1762 | |||
XM_017004626.1 | XP_016860115.1 | 14 | 1838 | |||
GenBank mRNA | AK127157.1 | - | 5 | 2193 | ||
AK225887.1 | - | 15 | 1460 | |||
AK297812.1 | - | 15 | 1551 | |||
BC012992.2 | - | 14 | 1487 | 3530897 | ||
BC012993.2 | - | 14 | 1487 | 3530898 | ||
BC018951.2 | - | 14 | 1788 | 4137722 | ||
BC051889.1 | - | 14 | 1374 | |||
BX648246.1 | - | 7 | 8958 | |||
L11289.1 | - | 14 | 1424 | |||
X64652.1 | - | 13 | 1307 | |||
X77494.1 | - | 16 | 1537 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
