siRNA Details
Gene Symbol: RMDN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170791.2 | NP_001164262.1 | 4, 5 | 856 | ||
NM_001170792.2 | NP_001164263.1 | 4, 5 | 927 | |||
NM_001170793.2 | NP_001164264.1 | 3, 4 | 433 | |||
NM_001322211.1 | NP_001309140.1 | 4, 5 | 925 | |||
NM_001322212.1 | NP_001309141.1 | 4, 5 | 856 | |||
NM_144713.4 | NP_653314.3 | 4, 5 | 1402 | |||
XM_011532613.2 | XP_011530915.1 | 4, 5 | 1383 | |||
XM_011532614.2 | XP_011530916.1 | 6, 7 | 1514 | |||
XM_011532615.2 | XP_011530917.2 | 6, 7 | 1514 | |||
XM_011532616.2 | XP_011530918.1 | 4, 5 | 828 | |||
XM_011532617.2 | XP_011530919.1 | 4, 5 | 914 | |||
XM_011532618.2 | XP_011530920.1 | 5, 6 | 1038 | |||
XM_011532619.2 | XP_011530921.1 | 4, 5 | 921 | |||
XM_017003474.1 | XP_016858963.1 | 6, 7 | 1514 | |||
XM_017003475.1 | XP_016858964.1 | 6, 7 | 1514 | |||
XM_017003476.1 | XP_016858965.1 | 4, 5 | 828 | |||
XM_017003477.1 | XP_016858966.1 | 4, 5 | 920 | |||
XM_017003478.1 | XP_016858967.1 | 4, 5 | 828 | |||
XM_017003479.1 | XP_016858968.1 | 6, 7 | 1007 | |||
GenBank mRNA | AF435956.1 | - | 3, 4 | 413 | ||
AK057516.1 | - | 4, 5 | 1384 | |||
AK095462.1 | - | 5, 6 | 913 | |||
AY358269.1 | - | 4, 5 | 856 | |||
BC024243.2 | - | 4, 5 | 929 | 4798767 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
