siRNA Details
Gene Symbol: GUCD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284251.1 | NP_001271180.1 | 6 | 2952 | ||
NM_001284252.1 | NP_001271181.1 | 6 | 2949 | |||
NM_001284253.1 | NP_001271182.1 | 6 | 2820 | |||
NM_001284254.1 | NP_001271183.1 | 6 | 2987 | |||
NM_001284255.1 | NP_001271184.1 | 6 | 2858 | |||
NM_001284256.1 | NP_001271185.1 | 5 | 2748 | |||
NM_001284257.1 | NP_001271186.1 | 5 | 2745 | |||
NM_031444.3 | NP_113632.2 | 6 | 2990 | |||
NR_104286.1 | 4 | 2625 | ||||
XM_005261761.1 | XP_005261818.1 | 5 | 2708 | |||
GenBank mRNA | AK024723.1 | - | 1 | 704 | ||
AK025242.1 | - | 1 | 1852 | |||
AK054681.1 | - | 6, 7 | 980 | |||
BC002924.2 | - | 5 | 2645 | |||
BC004144.1 | - | 1 | 501 | |||
BC053529.1 | - | 6 | 2644 | |||
BC070109.1 | - | 6 | 2993 | 4838028 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
