siRNA Details
Gene Symbol: GUCD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284251.1 | NP_001271180.1 | 6 | 3011 | ||
NM_001284252.1 | NP_001271181.1 | 6 | 3008 | |||
NM_001284253.1 | NP_001271182.1 | 6 | 2879 | |||
NM_001284254.1 | NP_001271183.1 | 6 | 3046 | |||
NM_001284255.1 | NP_001271184.1 | 6 | 2917 | |||
NM_001284256.1 | NP_001271185.1 | 5 | 2807 | |||
NM_001284257.1 | NP_001271186.1 | 5 | 2804 | |||
NM_031444.3 | NP_113632.2 | 6 | 3049 | |||
NR_104286.1 | 4 | 2684 | ||||
XM_005261761.1 | XP_005261818.1 | 5 | 2767 | |||
GenBank mRNA | AK024723.1 | - | 1 | 763 | ||
AK025242.1 | - | 1 | 1911 | |||
BC002924.2 | - | 5 | 2704 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
