siRNA Details
Gene Symbol: GUCD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284251.1 | NP_001271180.1 | 6 | 3163 | ||
NM_001284252.1 | NP_001271181.1 | 6 | 3160 | |||
NM_001284253.1 | NP_001271182.1 | 6 | 3031 | |||
NM_001284254.1 | NP_001271183.1 | 6 | 3198 | |||
NM_001284255.1 | NP_001271184.1 | 6 | 3069 | |||
NM_001284256.1 | NP_001271185.1 | 5 | 2959 | |||
NM_001284257.1 | NP_001271186.1 | 5 | 2956 | |||
NM_031444.3 | NP_113632.2 | 6 | 3201 | |||
NR_104286.1 | 4 | 2836 | ||||
XM_005261761.1 | XP_005261818.1 | 5 | 2919 | |||
GenBank mRNA | AK024723.1 | - | 1 | 915 | ||
AK025242.1 | - | 1 | 2063 | |||
AK054681.1 | - | 7 | 1191 | |||
BC002924.2 | - | 5 | 2856 | |||
BC004144.1 | - | 1 | 712 | |||
BC053529.1 | - | 6 | 2855 | |||
BC070109.1 | - | 6 | 3204 | 4838028 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
