siRNA Details
Gene Symbol: ATG10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001131028.1 | NP_001124500.1 | 3 | 457 | ||
NM_031482.4 | NP_113670.1 | 2 | 322 | |||
XM_005248610.4 | XP_005248667.1 | 2 | 1592 | |||
XM_005248611.4 | XP_005248668.1 | 2 | 166 | |||
XM_005248612.3 | XP_005248669.1 | 2 | 326 | |||
XM_017009944.1 | XP_016865433.1 | 3 | 461 | |||
GenBank mRNA | AF318326.1 | - | 2 | 475 | ||
AK024016.1 | - | 2 | 322 | |||
AK225969.1 | - | 2 | 322 | |||
AK315752.1 | - | 2 | 322 | |||
AL136912.1 | - | 2 | 190 | |||
BC029268.1 | - | 2 | 292 | 4521833 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
