siRNA Details
Gene Symbol: ATG10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001131028.1 | NP_001124500.1 | 4 | 609 | ||
NM_031482.4 | NP_113670.1 | 3 | 474 | |||
XM_005248610.4 | XP_005248667.1 | 3 | 1744 | |||
XM_005248611.4 | XP_005248668.1 | 3 | 318 | |||
XM_011543660.2 | XP_011541962.1 | 2 | 174 | |||
GenBank mRNA | AF318326.1 | - | 3 | 627 | ||
AK024016.1 | - | 3 | 474 | |||
AK315752.1 | - | 3 | 474 | |||
AL136912.1 | - | 3 | 342 | |||
BC029268.1 | - | 3 | 444 | 4521833 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
