siRNA Details
Gene Symbol: SEC31A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077206.3 | NP_001070674.1 | 26 | 3386 | ||
NM_001077207.3 | NP_001070675.1 | 27 | 3728 | |||
NM_001077208.3 | NP_001070676.1 | 27 | 3683 | |||
NM_001191049.2 | NP_001177978.1 | 26 | 3600 | |||
NM_001300744.2 | NP_001287673.1 | 24 | 3269 | |||
NM_001300745.2 | NP_001287674.1 | 25 | 3566 | |||
NM_001318119.1 | NP_001305048.1 | 27 | 3680 | |||
NM_001318120.1 | NP_001305049.1 | 29 | 3942 | |||
NM_016211.4 | NP_057295.2 | 25 | 3611 | |||
GenBank mRNA | AB018358.1 | - | 25 | 3467 | ||
AB018359.1 | - | 27 | 3584 | |||
AB020712.1 | - | 29 | 3909 | |||
AF139184.1 | - | 26 | 3531 | |||
AF161393.1 | - | 8 | 1017 | |||
AF161452.1 | - | 6 | 836 | |||
AK125897.1 | - | 20 | 3263 | |||
AK225934.1 | - | 27 | 3587 | |||
AK295810.1 | - | 26 | 3498 | |||
AL049463.1 | - | 1 | 26 | |||
AY137583.1 | - | 11 | 1395 | |||
BC117221.1 | - | 27 | 3551 | 40125772 | ||
BC143489.1 | - | 27 | 3548 | 9051997 | ||
BC143491.1 | - | 27 | 3551 | 9051999 | ||
BC143492.2 | - | 25 | 3434 | 9052000 | ||
CR933696.1 | - | 24 | 3142 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
