siRNA Details
Gene Symbol: SEC31A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077206.3 | NP_001070674.1 | 23 | 2899 | ||
NM_001077207.3 | NP_001070675.1 | 24 | 3241 | |||
NM_001077208.3 | NP_001070676.1 | 24 | 3196 | |||
NM_001191049.2 | NP_001177978.1 | 23 | 3113 | |||
NM_001300744.2 | NP_001287673.1 | 21 | 2782 | |||
NM_001300745.2 | NP_001287674.1 | 22 | 3079 | |||
NM_001318119.1 | NP_001305048.1 | 24 | 3193 | |||
NM_001318120.1 | NP_001305049.1 | 26 | 3455 | |||
NM_016211.4 | NP_057295.2 | 22 | 3124 | |||
GenBank mRNA | AB018358.1 | - | 22 | 2980 | ||
AB018359.1 | - | 24 | 3097 | |||
AB020712.1 | - | 26 | 3422 | |||
AF139184.1 | - | 23 | 3044 | |||
AF161393.1 | - | 5 | 531 | |||
AF161452.1 | - | 3 | 349 | |||
AK001014.1 | - | 13 | 1555 | |||
AK125897.1 | - | 17 | 2776 | |||
AK225934.1 | - | 24 | 3100 | |||
AK295810.1 | - | 23 | 3011 | |||
AY137583.1 | - | 8 | 908 | |||
BC084583.1 | - | 23 | 2738 | 6196402 | ||
BC117221.1 | - | 24 | 3064 | 40125772 | ||
BC143489.1 | - | 24 | 3061 | 9051997 | ||
BC143491.1 | - | 24 | 3064 | 9051999 | ||
BC143492.2 | - | 22 | 2947 | 9052000 | ||
CR933696.1 | - | 21 | 2655 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
