siRNA Details
Gene Symbol: SEC31A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077206.3 | NP_001070674.1 | 26 | 3432 | ||
NM_001077207.3 | NP_001070675.1 | 27 | 3774 | |||
NM_001077208.3 | NP_001070676.1 | 27 | 3729 | |||
NM_001191049.2 | NP_001177978.1 | 26 | 3646 | |||
NM_001300744.2 | NP_001287673.1 | 24 | 3315 | |||
NM_001300745.2 | NP_001287674.1 | 25 | 3612 | |||
NM_001318119.1 | NP_001305048.1 | 27 | 3726 | |||
NM_001318120.1 | NP_001305049.1 | 29 | 3988 | |||
NM_016211.4 | NP_057295.2 | 25 | 3657 | |||
GenBank mRNA | AB018358.1 | - | 25 | 3513 | ||
AB018359.1 | - | 27 | 3630 | |||
AB020712.1 | - | 29 | 3955 | |||
AF139184.1 | - | 26 | 3577 | |||
AF161393.1 | - | 8 | 1063 | |||
AF161452.1 | - | 6 | 882 | |||
AK125897.1 | - | 20 | 3309 | |||
AK225934.1 | - | 27 | 3633 | |||
AK295810.1 | - | 26 | 3544 | |||
AL049463.1 | - | 1 | 72 | |||
AY137583.1 | - | 11 | 1441 | |||
BC084583.1 | - | 26 | 3271 | 6196402 | ||
BC117221.1 | - | 27 | 3597 | 40125772 | ||
BC143489.1 | - | 27 | 3594 | 9051997 | ||
BC143491.1 | - | 27 | 3597 | 9051999 | ||
BC143492.2 | - | 25 | 3480 | 9052000 | ||
CR933696.1 | - | 24 | 3188 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
