siRNA Details
Gene Symbol: NCOA7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122842.2 | NP_001116314.1 | 16 | 3776 | ||
NM_001199619.1 | NP_001186548.1 | 17 | 3792 | |||
NM_001199620.1 | NP_001186549.1 | 18 | 3910 | |||
NM_001199621.1 | NP_001186550.1 | 15 | 3555 | |||
NM_001199622.1 | NP_001186551.1 | 6 | 1404 | |||
NM_181782.4 | NP_861447.3 | 16 | 3809 | |||
XM_005266822.4 | XP_005266879.1 | 16 | 3839 | |||
XM_006715340.3 | XP_006715403.1 | 16 | 3766 | |||
XM_011535455.2 | XP_011533757.1 | 16 | 3714 | |||
XM_017010269.1 | XP_016865758.1 | 16 | 3525 | |||
XM_017010270.1 | XP_016865759.1 | 17 | 3738 | |||
XM_017010271.1 | XP_016865760.1 | 16 | 3736 | |||
XM_017010272.1 | XP_016865761.1 | 16 | 3589 | |||
XM_017010273.1 | XP_016865762.1 | 16 | 3806 | |||
XM_017010274.1 | XP_016865763.1 | 16 | 3895 | |||
GenBank mRNA | AF493978.1 | - | 17 | 3792 | ||
AJ420542.1 | - | 8 | 1741 | |||
AK094706.1 | - | 6 | 1297 | |||
AL832621.2 | - | 16 | 3569 | |||
AL832628.2 | - | 16 | 3531 | |||
AL834442.1 | - | 14 | 3196 | |||
BC033612.1 | - | 9 | 2648 | |||
BX537385.1 | - | 16 | 3604 | |||
KC238672.1 | - | 6 | 1381 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
