siRNA Details
Gene Symbol: OR52B4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005161.3 | NP_001005161.2 | 1 | 962 | ||
GenBank mRNA | BC140928.1 | - | 1 | 962 | 9056740 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
