siRNA Details
Gene Symbol: TTC30B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152517.2 | NP_689730.2 | 1 | 2018 | ||
GenBank mRNA | AK055552.1 | - | 2 | 1917 | ||
AK296828.1 | - | 2 | 1853 | |||
BC033795.1 | - | 1 | 2002 | 5175299 | ||
BX647620.1 | - | 1 | 1551 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
