siRNA Details
Gene Symbol: TMEM135 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168724.1 | NP_001162195.1 | 3, 4 | 561 | ||
NM_022918.3 | NP_075069.3 | 3, 4 | 561 | |||
XM_017018140.1 | XP_016873629.1 | 2, 3 | 426 | |||
GenBank mRNA | AK025757.1 | - | 2, 3 | 175 | ||
AK225364.1 | - | 2, 3 | 175 | |||
AK291804.1 | - | 3, 4 | 471 | |||
BC030952.1 | - | 3, 4 | 420 | |||
BC051462.1 | - | 3, 4 | 382 | 5278478 | ||
BX648678.1 | - | 3, 4 | 547 |

More Information
Other Species Targeted

Gene Ontology Categories:
