siRNA Details
Gene Symbol: LOC400661 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_122073.1 | 1 | 385 | |||
GenBank mRNA | AK126829.1 | - | 1 | 385 | ||
BC119774.1 | - | 1 | 325 | 40109466 | ||
BC119776.1 | - | 1 | 325 | 40109460 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
