siRNA Details
Gene Symbol: SRCIN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025248.2 | NP_079524.2 | 12, 13 | 2556 | ||
XM_017025169.1 | XP_016880658.1 | 13, 14 | 2868 | |||
XM_017025170.1 | XP_016880659.1 | 13, 14 | 2723 | |||
XM_017025171.1 | XP_016880660.1 | 13, 14 | 2723 | |||
XM_017025172.1 | XP_016880661.1 | 12, 13 | 2621 | |||
XM_017025173.1 | XP_016880662.1 | 13, 14 | 2723 | |||
XM_017025174.1 | XP_016880663.1 | 11, 12 | 2163 | |||
GenBank mRNA | AB051471.1 | - | 6, 7 | 1068 | ||
AB127405.1 | - | 12, 13 | 2556 | |||
AK126665.1 | - | 8, 9 | 2266 | |||
AK126676.1 | - | 8, 9 | 1693 | |||
AK295027.1 | - | 11, 12 | 2163 | |||
BC033233.1 | - | 3, 4 | 713 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
