siRNA Details
Gene Symbol: SRCIN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025248.2 | NP_079524.2 | 12 | 2469 | ||
XM_017025169.1 | XP_016880658.1 | 13 | 2781 | |||
XM_017025170.1 | XP_016880659.1 | 13 | 2636 | |||
XM_017025171.1 | XP_016880660.1 | 13 | 2636 | |||
XM_017025172.1 | XP_016880661.1 | 12 | 2534 | |||
XM_017025173.1 | XP_016880662.1 | 13 | 2636 | |||
XM_017025174.1 | XP_016880663.1 | 11 | 2076 | |||
GenBank mRNA | AB051471.1 | - | 6 | 981 | ||
AB127405.1 | - | 12 | 2469 | |||
AK126665.1 | - | 8 | 2179 | |||
AK126676.1 | - | 8 | 1606 | |||
AK295027.1 | - | 11 | 2076 | |||
BC033233.1 | - | 3 | 626 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
