siRNA Details
Gene Symbol: MYO1D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303279.1 | NP_001290208.1 | 9 | 1421 | ||
NM_001303280.1 | NP_001290209.1 | 9 | 1421 | |||
NM_015194.2 | NP_056009.1 | 9 | 1421 | |||
XM_017024685.1 | XP_016880174.1 | 9 | 1392 | |||
GenBank mRNA | AB018270.2 | - | 9 | 1132 | ||
AK026920.1 | - | 6 | 717 | |||
AK127942.1 | - | 9 | 1392 | |||
BC146763.1 | - | 9 | 1132 | 9007203 | ||
BX537615.1 | - | 8 | 854 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
