siRNA Details
Gene Symbol: LINC01565 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125802.1 | 3 | 1358 | |||
GenBank mRNA | AF008192.1 | - | 3 | 1358 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
