siRNA Details
Gene Symbol: CASS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164114.1 | NP_001157586.1 | 3 | 526 | ||
NM_001164115.1 | NP_001157587.1 | 3 | 688 | |||
NM_001164116.1 | NP_001157588.1 | 4 | 712 | |||
NM_020356.3 | NP_065089.2 | 3 | 688 | |||
XM_006723831.3 | XP_006723894.1 | 4 | 1112 | |||
GenBank mRNA | AJ276678.1 | - | 4 | 712 | ||
AK027760.1 | - | 3 | 685 | |||
AK295613.1 | - | 3 | 391 | |||
AK308057.1 | - | 3 | 689 | |||
AK314653.1 | - | 3 | 621 | |||
BC027951.1 | - | 3 | 638 | 5205865 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
