siRNA Details
Gene Symbol: OTOP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178160.2 | NP_835454.1 | 6 | 1308 | ||
XM_011525479.1 | XP_011523781.1 | 7 | 1518 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
