siRNA Details
Gene Symbol: OTOP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178160.2 | NP_835454.1 | 2 | 240 | ||
XM_011525479.1 | XP_011523781.1 | 3 | 450 | |||
GenBank mRNA | AK000063.1 | - | 2 | 240 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
