siRNA Details
Gene Symbol: LEKR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004316.2 | NP_001004316.2 | 10 | 1240 | ||
XM_005247441.3 | XP_005247498.1 | 10 | 3052 | |||
XM_006713631.3 | XP_006713694.1 | 10 | 1764 | |||
XM_006713636.2 | XP_006713699.1 | 5 | 460 | |||
XM_011512803.2 | XP_011511105.1 | 10 | 1764 | |||
XM_011512804.2 | XP_011511106.1 | 10 | 3285 | |||
XM_011512809.1 | XP_011511111.1 | 7 | 871 | |||
XM_011512813.2 | XP_011511115.1 | 5 | 544 | |||
XM_011512814.2 | XP_011511116.1 | 5 | 452 | |||
XM_017006374.1 | XP_016861863.1 | 6 | 648 | |||
XM_017006375.1 | XP_016861864.1 | 6 | 639 | |||
GenBank mRNA | AK131473.1 | - | 11 | 1549 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
