siRNA Details
Gene Symbol: TPRG1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182752.3 | NP_877429.2 | 5 | 738 | ||
GenBank mRNA | AK025464.1 | - | 4 | 458 | ||
AK096427.1 | - | 5 | 715 | |||
AK289919.1 | - | 5 | 688 | |||
AK298343.1 | - | 5 | 577 | |||
BC019034.2 | - | 4 | 503 | 4576259 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
