siRNA Details
Gene Symbol: CXorf23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198279.3 | NP_938020.2 | 7 | 1645 | ||
XM_005274473.3 | XP_005274530.1 | 8 | 2202 | |||
XM_005274474.3 | XP_005274531.1 | 8 | 2142 | |||
XM_005274475.3 | XP_005274532.1 | 8 | 2202 | |||
XM_011545475.2 | XP_011543777.1 | 8 | 1812 | |||
XM_011545476.2 | XP_011543778.1 | 8 | 1779 | |||
XM_011545477.2 | XP_011543779.1 | 8 | 2200 | |||
XM_011545478.2 | XP_011543780.1 | 8 | 2202 | |||
XM_011545479.2 | XP_011543781.1 | 8 | 2201 | |||
XM_011545482.2 | XP_011543784.1 | 8 | 2200 | |||
XM_017029390.1 | XP_016884879.1 | 8 | 2140 | |||
XM_017029391.1 | XP_016884880.1 | 8 | 2142 | |||
GenBank mRNA | AK094661.1 | - | 5 | 470 | ||
AK127709.1 | - | 6 | 1261 | |||
AL833278.1 | - | 6 | 1495 | |||
BC126172.1 | - | 5 | 780 | |||
BC143906.1 | - | 5 | 780 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
