siRNA Details
Gene Symbol: THEM5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182578.3 | NP_872384.1 | 4 | 630 | ||
XM_011509421.1 | XP_011507723.1 | 4 | 630 | |||
GenBank mRNA | AK095283.1 | - | 4 | 630 | ||
BC101610.1 | - | 4 | 536 | 8069116 | ||
BC112239.1 | - | 4 | 536 | 8327707 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
