siRNA Details
Gene Symbol: SAP130 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145928.1 | NP_001139400.1 | 13 | 1774 | ||
NM_024545.3 | NP_078821.2 | 13 | 1774 | |||
XM_005263767.3 | XP_005263824.1 | 13 | 1745 | |||
XM_005263768.3 | XP_005263825.1 | 13 | 1724 | |||
XM_005263769.3 | XP_005263826.1 | 13 | 1645 | |||
XM_005263770.3 | XP_005263827.1 | 13 | 1721 | |||
XM_006712748.3 | XP_006712811.1 | 13 | 1934 | |||
XM_006712749.3 | XP_006712812.1 | 13 | 1934 | |||
XM_017004903.1 | XP_016860392.1 | 13 | 1724 | |||
XM_017004904.1 | XP_016860393.1 | 13 | 1724 | |||
XM_017004905.1 | XP_016860394.1 | 13 | 1724 | |||
GenBank mRNA | AK022823.1 | - | 3 | 291 | ||
AK055979.1 | - | 13 | 1687 | |||
AK092234.1 | - | 7 | 835 | |||
AK315185.1 | - | 13 | 1774 | |||
AL136833.1 | - | 13 | 1772 | |||
AY220791.1 | - | 13 | 1642 | |||
BC017453.1 | - | 6 | 579 | |||
BC117255.1 | - | 13 | 1740 | 40125806 | ||
BC143898.1 | - | 13 | 1740 | 9052416 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
