siRNA Details
Gene Symbol: LINC00477 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_029451.2 | 1 | 433 | |||
GenBank mRNA | AK057456.1 | - | 1 | 433 | ||
BC104436.1 | - | 1 | 413 | |||
BC104437.1 | - | 1 | 413 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
