siRNA Details
Gene Symbol: ANKRD46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270377.1 | NP_001257306.1 | 4 | 583 | ||
NM_001270378.1 | NP_001257307.1 | 4 | 1181 | |||
NM_001270379.1 | NP_001257308.1 | 4 | 583 | |||
NM_198401.3 | NP_940683.1 | 5 | 688 | |||
XM_006716509.1 | XP_006716572.1 | 4 | 724 | |||
XM_017013106.1 | XP_016868595.1 | 4 | 646 | |||
GenBank mRNA | AB077205.1 | - | 5 | 686 | ||
AK095235.1 | - | 4 | 558 | |||
AL832842.1 | - | 4 | 543 | |||
BC050396.1 | - | 4 | 539 | 6065488 | ||
BC060843.1 | - | 4 | 556 | 30341860 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
