siRNA Details
Gene Symbol: GSE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134473.2 | NP_001127945.1 | 14, 15 | 3413 | ||
NM_001278184.1 | NP_001265113.1 | 14, 15 | 3467 | |||
NM_014615.3 | NP_055430.1 | 15, 16 | 3686 | |||
XM_005255859.4 | XP_005255916.3 | 16, 17 | 5828 | |||
XM_005255860.3 | XP_005255917.3 | 15, 16 | 5721 | |||
XM_005255861.4 | XP_005255918.3 | 15, 16 | 5609 | |||
XM_005255863.4 | XP_005255920.1 | 15, 16 | 3578 | |||
XM_005255864.4 | XP_005255921.1 | 15, 16 | 4310 | |||
XM_005255865.4 | XP_005255922.1 | 14, 15 | 4091 | |||
XM_011522965.2 | XP_011521267.1 | 15, 16 | 4077 | |||
XM_011522966.2 | XP_011521268.1 | 15, 16 | 3544 | |||
XM_017023083.1 | XP_016878572.1 | 15, 16 | 3836 | |||
GenBank mRNA | AK127344.1 | - | 4, 5 | 2097 | ||
AK308612.1 | - | 6, 7 | 1263 | |||
BC000753.2 | - | 3, 4 | 655 | |||
BC009854.2 | - | 2, 3 | 160 | |||
BC037556.1 | - | 14, 15 | 3436 | 4994198 | ||
D80004.2 | - | 14, 15 | 3351 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
