siRNA Details
Gene Symbol: GSE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134473.2 | NP_001127945.1 | 4 | 535 | ||
NM_001278184.1 | NP_001265113.1 | 4 | 589 | |||
NM_014615.3 | NP_055430.1 | 5 | 808 | |||
XM_005255859.4 | XP_005255916.3 | 6 | 2950 | |||
XM_005255860.3 | XP_005255917.3 | 6 | 2957 | |||
XM_005255861.4 | XP_005255918.3 | 5 | 2731 | |||
XM_005255863.4 | XP_005255920.1 | 5 | 700 | |||
XM_005255864.4 | XP_005255921.1 | 5 | 1432 | |||
XM_005255865.4 | XP_005255922.1 | 4 | 1213 | |||
XM_011522965.2 | XP_011521267.1 | 5 | 1199 | |||
XM_011522966.2 | XP_011521268.1 | 5 | 666 | |||
XM_017023083.1 | XP_016878572.1 | 5 | 958 | |||
GenBank mRNA | BC037556.1 | - | 4 | 558 | 4994198 | |
D80004.2 | - | 4 | 473 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
