siRNA Details
Gene Symbol: GAS2L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139285.3 | NP_644814.1 | 3 | 680 | ||
GenBank mRNA | AF508784.1 | - | 4 | 633 | ||
AF508785.1 | - | 3 | 680 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
