siRNA Details
Gene Symbol: AP4M1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004722.3 | NP_004713.2 | 3 | 411 | ||
XM_005250689.4 | XP_005250746.1 | 4 | 315 | |||
XM_006716175.3 | XP_006716238.1 | 3 | 393 | |||
XM_011516685.2 | XP_011514987.1 | 3 | 393 | |||
XM_017012790.1 | XP_016868279.1 | 2 | 389 | |||
GenBank mRNA | AF020796.1 | - | 3 | 362 | ||
AF155158.1 | - | 3 | 247 | |||
AK055686.1 | - | 2 | 388 | |||
AK293590.1 | - | 2 | 180 | |||
AK296651.1 | - | 2 | 167 | |||
AK304489.1 | - | 3 | 241 | |||
AK315782.1 | - | 3 | 228 | |||
BC018705.1 | - | 3 | 254 | 5013882 | ||
BX640759.1 | - | 2 | 160 | |||
DC413622.1 | - | 3 | 411 | |||
Y08387.1 | - | 3 | 257 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
