siRNA Details
Gene Symbol: SLC39A14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128431.2 | NP_001121903.1 | 6 | 950 | ||
NM_001135153.1 | NP_001128625.1 | 6 | 991 | |||
NM_001135154.1 | NP_001128626.1 | 6 | 950 | |||
NM_015359.4 | NP_056174.2 | 6 | 950 | |||
XM_005273465.2 | XP_005273522.1 | 7 | 1038 | |||
XM_005273466.4 | XP_005273523.1 | 6 | 1022 | |||
XM_006716323.2 | XP_006716386.1 | 8 | 1236 | |||
XM_006716324.2 | XP_006716387.1 | 7 | 1027 | |||
XM_011544478.2 | XP_011542780.1 | 7 | 1102 | |||
XM_017013292.1 | XP_016868781.1 | 7 | 1118 | |||
XM_017013293.1 | XP_016868782.1 | 8 | 1236 | |||
XM_017013294.1 | XP_016868783.1 | 8 | 1145 | |||
XM_017013295.1 | XP_016868784.1 | 6 | 1155 | |||
GenBank mRNA | AK172810.1 | - | 6 | 921 | ||
AK295807.1 | - | 6 | 866 | |||
BC015770.1 | - | 6 | 912 | 4865469 | ||
D31887.1 | - | 6 | 894 | |||
KU178568.1 | - | 5 | 775 | |||
KU178569.1 | - | 5 | 775 | |||
KU178570.1 | - | 5 | 775 | |||
KU178571.1 | - | 5 | 775 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
