siRNA Details
Gene Symbol: ZNF701 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172655.1 | NP_001166126.1 | 5 | 765 | ||
NM_018260.2 | NP_060730.2 | 4 | 471 | |||
XM_011527092.1 | XP_011525394.1 | 4 | 545 | |||
GenBank mRNA | BC054884.1 | - | 4 | 439 | ||
BC067346.1 | - | 5 | 765 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
