siRNA Details
Gene Symbol: ZNF701 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172655.1 | NP_001166126.1 | 5 | 742 | ||
NM_018260.2 | NP_060730.2 | 4 | 448 | |||
XM_011527092.1 | XP_011525394.1 | 4 | 522 | |||
GenBank mRNA | AK001753.1 | - | 4 | 446 | ||
BC054884.1 | - | 4 | 416 | |||
BC067346.1 | - | 5 | 742 | |||
BC080612.1 | - | 3 | 404 | 5816672 | ||
BC131719.1 | - | 4 | 422 | 40123413 | ||
BC136425.1 | - | 4 | 552 | 9020414 | ||
BC136426.1 | - | 4 | 420 | 9020415 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
